Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Difluorobenzophenone, 97+%

Click to view available options
Quantity:
1g
5g
25g
85068-36-6
C13H8F2O
218.203
MFCD00009899
HSCUAAMDKDZZKG-UHFFFAOYSA-N
2,5-difluorobenzophenone,2,5-difluorophenyl phenyl methanone,2,5-difluoro-phenyl-phenyl-methanone,methanone, 2,5-difluorophenyl phenyl,2,5-difluoro diphenylmethanone,2,5-difluorophenyl-phenylmethanone,2,5-difluorophenyl phenyl ketone,pubchem4214,2.5-difluorobenzophenone,2,5-difluoro benzophenone
522826
(2,5-difluorophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)F)F
85068-36-6
C13H8F2O
218.203
MFCD00009899
HSCUAAMDKDZZKG-UHFFFAOYSA-N
2,5-difluorobenzophenone,2,5-difluorophenyl phenyl methanone,2,5-difluoro-phenyl-phenyl-methanone,methanone, 2,5-difluorophenyl phenyl,2,5-difluoro diphenylmethanone,2,5-difluorophenyl-phenylmethanone,2,5-difluorophenyl phenyl ketone,pubchem4214,2.5-difluorobenzophenone,2,5-difluoro benzophenone
522826
(2,5-difluorophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=C(C=CC(=C2)F)F
Refractive Index 1.5693
Density 1.26
Flash Point >110°C (230°F)
Quantity 25g
Boiling Point 120°C to 123°C (0.4mmHg)
Beilstein 4417185
Formula Weight 218.2
Percent Purity ≥97%
Chemical Name or Material 2,5-Difluorobenzophenone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 285-298-2

TSCA : No

Recommended Storage : Ambient temperatures