Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluorobenzyl Alcohol 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
75853-20-2
C7H6F2O
144.121
MFCD00010627
KIKCAPPVFQLOIU-UHFFFAOYSA-N
2,5-difluorobenzyl alcohol,2,5-difluorophenyl methanol,benzenemethanol, 2,5-difluoro,2,5-difluorobenzylalcohol,2,5-difluorophenyl methan-1-ol,pubchem4913,acmc-209p0i,rarechem al bd 0214,2,5-difluoro-phenyl-methanol
522599
(2,5-difluorophenyl)methanol
C1=CC(=C(C=C1F)CO)F
75853-20-2
C7H6F2O
144.121
MFCD00010627
KIKCAPPVFQLOIU-UHFFFAOYSA-N
2,5-difluorobenzyl alcohol,2,5-difluorophenyl methanol,benzenemethanol, 2,5-difluoro,2,5-difluorobenzylalcohol,2,5-difluorophenyl methan-1-ol,pubchem4913,acmc-209p0i,rarechem al bd 0214,2,5-difluoro-phenyl-methanol
522599
(2,5-difluorophenyl)methanol
C1=CC(=C(C=C1F)CO)F
Quantity 25g
Color Colorless
Formula Weight 144.12
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,5-Difluorobenzyl Alcohol