Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluorobenzyl Chloride, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
495-07-8
C7H5ClF2
162.564
MFCD01090989
INXKTZMJFPRVAY-UHFFFAOYSA-N
2,5-difluorobenzyl chloride,2-chloromethyl-1,4-difluorobenzene,2,5-difluorobenzylchloride,alpha-chloro-2,5-difluorotoluene,benzene, 2-chloromethyl-1,4-difluoro,2-chloromethyl-1,4-bis fluoranyl benzene,2-chloromethyl-1,4-difluoro-benzene,acmc-1an3f,ksc591g4f
2736950
2-(chloromethyl)-1,4-difluorobenzene
C1=CC(=C(C=C1F)CCl)F
495-07-8
C7H5ClF2
162.564
MFCD01090989
INXKTZMJFPRVAY-UHFFFAOYSA-N
2,5-difluorobenzyl chloride,2-chloromethyl-1,4-difluorobenzene,2,5-difluorobenzylchloride,alpha-chloro-2,5-difluorotoluene,benzene, 2-chloromethyl-1,4-difluoro,2-chloromethyl-1,4-bis fluoranyl benzene,2-chloromethyl-1,4-difluoro-benzene,acmc-1an3f,ksc591g4f
2736950
2-(chloromethyl)-1,4-difluorobenzene
C1=CC(=C(C=C1F)CCl)F
Infrared Spectrum Authentic
Refractive Index 1.4920 to 1.4940 (20°C, 589nm)
Packaging Glass bottle
Color Colorless
Quantity 25g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula ClCH2C6H3F2
Formula Weight 162.57
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2, 5-Difluorobenzyl chloride
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger