Alfa Aesar™ 2,5-Difluorothiobenzamide, 97%

Manufacturer: Alfa Aesar™ H3222503

Quantity 1g

 View more versions of this product

Catalog No. AAH3222503

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C7H5F2NS
    Formula Weight 173.18
    MDL Number MFCD09757552
    UN Number UN2811
    Chemical Name or Material 2,5-Difluorothiobenzamide

    Specifications

    Formula Weight 173.18
    Molecular Formula C7H5F2NS
    MDL Number MFCD09757552
    UN Number UN2811
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Health Hazard 3 GHS P Statement
    P280f-P309-P310
    Wear protective clothing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    Melting Point 109° to 113°C
    Quantity 1g
    Chemical Name or Material 2,5-Difluorothiobenzamide
    Assay 97%

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P280f-P309-P310
    Wear protective clothing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit