Alfa Aesar™ 2,5-Difluorotoluene, 98%

Manufacturer: Alfa Aesar™ B2254406

Quantity 5g

 View more versions of this product

Catalog No. AAB2254406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 452-67-5
    Molecular Formula C7H6F2
    Formula Weight 128.12
    MDL Number MFCD00000347
    UN Number UN2388
    Beilstein 2041492
    Chemical Name or Material 2,5-Difluorotoluene

    Specifications

    Formula Weight 128.12
    Molecular Formula C7H6F2
    MDL Number MFCD00000347
    CAS 452-67-5
    UN Number UN2388
    Beilstein 2041492
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROTOLUENES
    Flash Point 12°C (53°F)
    Odor Aromatic
    Boiling Point 115° to 117°C
    Melting Point −35°C
    Quantity 5g
    Chemical Name or Material 2,5-Difluorotoluene
    Assay 98%

    GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit