Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Difluorotoluene, 98%

Click to view available options
Quantity:
5g
25g
452-67-5
C7H6F2
128.122
MFCD00000347
YSNVKDGEALPJGC-UHFFFAOYSA-N
2,5-difluorotoluene,benzene, 1,4-difluoro-2-methyl,1,4-difluoro-2-methyl-benzene,1,4-difluor-2-methylbenzol,2,5-difluorotolune,2,5-difluoro toluene,p-ditrifluorotoluene,acmc-209k1g,ksc236k9r,benzene,4-difluoro-2-methyl
67980
1,4-difluoro-2-methylbenzene
CC1=C(C=CC(=C1)F)F
452-67-5
C7H6F2
128.122
MFCD00000347
YSNVKDGEALPJGC-UHFFFAOYSA-N
2,5-difluorotoluene,benzene, 1,4-difluoro-2-methyl,1,4-difluoro-2-methyl-benzene,1,4-difluor-2-methylbenzol,2,5-difluorotolune,2,5-difluoro toluene,p-ditrifluorotoluene,acmc-209k1g,ksc236k9r,benzene,4-difluoro-2-methyl
67980
1,4-difluoro-2-methylbenzene
CC1=C(C=CC(=C1)F)F
Melting Point -35°C
Boiling Point 115°C to 117°C
Quantity 25g
Flash Point 12°C (53°F)
Odor Aromatic
UN Number UN2388
Beilstein 2041492
Formula Weight 128.12
Percent Purity 98%
Chemical Name or Material 2,5-Difluorotoluene

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROTOLUENES