Alfa Aesar™ 2,5-Dihydro-2,5-dimethoxyfuran, cis + trans, 99%

Manufacturer: Alfa Aesar™ B2502836

Quantity 500g

 View more versions of this product

Catalog No. AAB2502836

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 332-77-4
    Molecular Formula C6H10O3
    Formula Weight 130.14
    MDL Number MFCD00003220
    UN Number UN3271
    Beilstein 113225
    Synonym 2,5-Dimethoxy-2,5-dihydrofuran
    Chemical Name or Material 2,5-Dihydro-2,5-dimethoxyfuran, cis + trans

    Specifications

    Formula Weight 130.14
    MDL Number MFCD00003220
    CAS 332-77-4
    UN Number UN3271
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H226-H332
    Toxic if swallowed.
    Flammable liquid and vapour.
    Harmful if inhaled.
    Health Hazard 3 GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
    TSCA TSCA
    Molecular Formula C6H10O3
    Melting Point <−40°C
    Beilstein 113225
    Synonym 2,5-Dimethoxy-2,5-dihydrofuran
    Flash Point 47°C (116°F)
    Boiling Point 160° to 162°C
    Quantity 500g
    Chemical Name or Material 2,5-Dihydro-2,5-dimethoxyfuran, cis + trans
    Assay 99%

    GHS H Statement
    H301-H226-H332
    Toxic if swallowed.
    Flammable liquid and vapour.
    Harmful if inhaled.

    GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit