Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dihydro-2,5-dimethoxyfuran, cis + trans, 99%, Thermo Scientific Chemicals

Catalog Number AAB2502830
Click to view available options
Quantity:
100 g
250 g
500 g
1000 g
332-77-4
C6H10O3
130.14
MFCD00003220
WXFWXFIWDGJRSC-UHFFFAOYNA-N
2,5-dihydro-2,5-dimethoxyfuran, furan, 2,5-dihydro-2,5-dimethoxy, 2,5-dihydro-2,5-dimethoxyfuran, cis+trans, 2,5-dihydro-2,5-dimethoxyfuran, mixture of cis and trans, pubchem6933, ksc490q3d, furan,5-dihydro-2,5-dimethoxy, 2,5-dihydro-2,5-dimethoxyfuran,c&t, 2,5-dimethoxy-2,5-dihydrofuran,c&t, 2,5-dihydro-2,5-dimethoxyfuran, cis + trans
78974
2,5-dimethoxy-2,5-dihydrofuran
COC1OC(OC)C=C1
332-77-4
C6H10O3
130.14
MFCD00003220
WXFWXFIWDGJRSC-UHFFFAOYNA-N
2,5-dihydro-2,5-dimethoxyfuran, furan, 2,5-dihydro-2,5-dimethoxy, 2,5-dihydro-2,5-dimethoxyfuran, cis+trans, 2,5-dihydro-2,5-dimethoxyfuran, mixture of cis and trans, pubchem6933, ksc490q3d, furan,5-dihydro-2,5-dimethoxy, 2,5-dihydro-2,5-dimethoxyfuran,c&t, 2,5-dimethoxy-2,5-dihydrofuran,c&t, 2,5-dihydro-2,5-dimethoxyfuran, cis + trans
78974
2,5-dimethoxy-2,5-dihydrofuran
COC1OC(OC)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point <-40°C
Density 1.072
Boiling Point 160°C to 162°C
Flash Point 47°C (116°F)
Refractive Index 1.435
Quantity 250 g
UN Number UN3271
Beilstein 113225
Formula Weight 130.14
Percent Purity 99%
Chemical Name or Material 2,5-Dihydro-2,5-dimethoxyfuran, cis + trans
Show More Show Less
Hazard Category H226-H302+H332-H319
Hazard Statement GHS H Statement
H301-H226-H332
Toxic if swallowed.
Flammable liquid and vapour.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 206-367-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit