Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Dihydro-2,5-dimethoxyfuran, cis + trans, 99%

Click to view available options
Quantity:
100g
250g
500g
1000g
332-77-4
C6H10O3
130.143
MFCD00003220
WXFWXFIWDGJRSC-UHFFFAOYSA-N
2,5-dihydro-2,5-dimethoxyfuran,furan, 2,5-dihydro-2,5-dimethoxy,2,5-dihydro-2,5-dimethoxyfuran, cis+trans,2,5-dihydro-2,5-dimethoxyfuran, mixture of cis and trans,pubchem6933,ksc490q3d,furan,5-dihydro-2,5-dimethoxy,2,5-dihydro-2,5-dimethoxyfuran,c&t,2,5-dimethoxy-2,5-dihydrofuran,c&t,2,5-dihydro-2,5-dimethoxyfuran, cis + trans
78974
2,5-dimethoxy-2,5-dihydrofuran
COC1C=CC(O1)OC
332-77-4
C6H10O3
130.143
MFCD00003220
WXFWXFIWDGJRSC-UHFFFAOYSA-N
2,5-dihydro-2,5-dimethoxyfuran,furan, 2,5-dihydro-2,5-dimethoxy,2,5-dihydro-2,5-dimethoxyfuran, cis+trans,2,5-dihydro-2,5-dimethoxyfuran, mixture of cis and trans,pubchem6933,ksc490q3d,furan,5-dihydro-2,5-dimethoxy,2,5-dihydro-2,5-dimethoxyfuran,c&t,2,5-dimethoxy-2,5-dihydrofuran,c&t,2,5-dihydro-2,5-dimethoxyfuran, cis + trans
78974
2,5-dimethoxy-2,5-dihydrofuran
COC1C=CC(O1)OC
Melting Point <-40°C
Boiling Point 160°C to 162°C
Flash Point 47°C (116°F)
Quantity 100g
UN Number UN3271
Beilstein 113225
Formula Weight 130.14
Percent Purity 99%
Chemical Name or Material 2,5-Dihydro-2,5-dimethoxyfuran, cis + trans

GHS H Statement
H301-H226-H332
Toxic if swallowed.
Flammable liquid and vapour.
Harmful if inhaled.

GHS P Statement
P210-P261-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.

TSCA : TSCA