Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dihydroxy-1,4-benzoquinone, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
615-94-1
C6H4O4
140.1
MFCD00001598
QFSYADJLNBHAKO-UHFFFAOYSA-N
2,5-dihydroxy-1,4-benzoquinone,2,5-dihydroxy-p-benzoquinone,2,5-cyclohexadiene-1,4-dione, 2,5-dihydroxy,p-benzoquinone, 2,5-dihydroxy,2,5-dihydroxybenzoquinone,4lbp,2,5-dihydroxy-p-quinone,2,5-dihydroxy benzoquinone,2,5-dhbqop
69213
2,5-dihydroxycyclohexa-2,5-diene-1,4-dione
C1=C(C(=O)C=C(C1=O)O)O
615-94-1
C6H4O4
140.1
MFCD00001598
QFSYADJLNBHAKO-UHFFFAOYSA-N
2,5-dihydroxy-1,4-benzoquinone,2,5-dihydroxy-p-benzoquinone,2,5-cyclohexadiene-1,4-dione, 2,5-dihydroxy,p-benzoquinone, 2,5-dihydroxy,2,5-dihydroxybenzoquinone,4lbp,2,5-dihydroxy-p-quinone,2,5-dihydroxy benzoquinone,2,5-dhbqop
69213
2,5-dihydroxycyclohexa-2,5-diene-1,4-dione
C1=C(C(=O)C=C(C1=O)O)O
Melting Point 220°C
Quantity 100g
Color Ochre to Brown
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula (HO)2C6H2(=O)2
Beilstein 08, 377
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,acetic acid;,slightly soluble in acetone,dmso;,insoluble in diethyl ether
Formula Weight 140.1
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2, 5-Dihydroxy-1, 4-benzoquinone, 98%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-454-3

TSCA : TSCA