Learn More
2,5-Dihydroxybenzoic acid, 99%, Thermo Scientific Chemicals
Description
2,5-dihydroxybenzoic acid are used as intermediates for pharmaceuticals (especially for antipyetic anlgesic, antirheumatism) and other organic synthesis. They are used as matrix for ionization of peptides, proteins and carbohydrates. In industrial field, they are used as intermediates for the production of other organic chemicals, resins, polyesters, plasticizers, dyestuffs, preservatives, and rubber chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,5-dihydroxybenzoic acid are used as intermediates for pharmaceuticals (especially for antipyetic anlgesic, antirheumatism) and other organic synthesis. They are used as matrix for ionization of peptides, proteins and carbohydrates. In industrial field, they are used as intermediates for the production of other organic chemicals, resins, polyesters, plasticizers, dyestuffs, preservatives, and rubber chemicals.
Solubility
It is soluble in alcohol and ether.
Notes
Keep away from Strong oxidizing agents, Strong bases, Acid chlorides, Acid anhydrides. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | ∼206°C (decomposition) |
Quantity | 25 g |
Beilstein | 2209119 |
Merck Index | 14,4398 |
Solubility Information | It is soluble in alcohol and ether. |
Formula Weight | 154.12 |
Percent Purity | 99% |
Chemical Name or Material | 2,5-Dihydroxybenzoic acid |
Safety and Handling
Hazard Category | H302-H315-H319-H335-H500 |
Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 207-718-5 |
RTECSNumber | LY3850000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.