Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimercapto-1,3,4-thiadiazole 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Plastic bottle
1072-71-5
C2H2N2S3
150.23
MFCD00003103
BIGYLAKFCGVRAN-UHFFFAOYSA-N
1,3,4-thiadiazole-2,5-dithiol,2,5-dimercapto-1,3,4-thiadiazole,bismuthiol i,dimercaptothiadiazole,bismuththiol,2,5-dimercaptothiadiazole,usaf a-8354,bismuthiol,py 61h,bismuththiol i
2723630
1,3,4-thiadiazolidine-2,5-dithione
C1(=S)NNC(=S)S1
1072-71-5
C2H2N2S3
150.23
MFCD00003103
BIGYLAKFCGVRAN-UHFFFAOYSA-N
1,3,4-thiadiazole-2,5-dithiol,2,5-dimercapto-1,3,4-thiadiazole,bismuthiol i,dimercaptothiadiazole,bismuththiol,2,5-dimercaptothiadiazole,usaf a-8354,bismuthiol,py 61h,bismuththiol i
2723630
1,3,4-thiadiazolidine-2,5-dithione
C1(=S)NNC(=S)S1
Melting Point 161°C to 165°C
Packaging Plastic bottle
Infrared Spectrum Authentic
Quantity 100g
Color White to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 27, 677
Solubility Information Solubility in water: 20g/L water (20°C)
Formula Weight 150.23
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2,5-Dimercapto-1,3,4-thiadiazole, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

STENCH

EINECSNumber : 214-014-1

RTECSNumber : XI3850000

TSCA : TSCA