Learn More
2,5-Dimercapto-1,3,4-thiadiazole dipotassium salt, 97%, Thermo Scientific Chemicals
Description
1,3,4-thiadiazole-2,5-dithiol (Bismuthiol I) was used in determination of tellurium in indium phosphide doped with cadmium telluride by electro thermal atomic absorption spectrometry. It forms polymers with metals and can be useful in treatment of metal-polluted water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,3,4-thiadiazole-2,5-dithiol (Bismuthiol I) was used in determination of tellurium in indium phosphide doped with cadmium telluride by electro thermal atomic absorption spectrometry. It forms polymers with metals and can be useful in treatment of metal-polluted water.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | ∽274°C (decomposition) |
Quantity | 5 g |
Beilstein | 4917786 |
Solubility Information | Insoluble in water. |
Formula Weight | 226.43 |
Percent Purity | 97% |
Chemical Name or Material | 2,5-Dimercapto-1,3,4-thiadiazole dipotassium salt |
Safety and Handling
EINECSNumber | 225-043-4 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.