Learn More
2,5-Dimethoxy-3-tetrahydrofurancarboxaldehyde, 90%, Tech., mixture of isomers, ACROS Organics™
Specifications
Color | Yellow to Brown |
Density | 1.129g/mL |
Infrared Spectrum | Authentic |
Quantity | 5g |
Packaging | Glass bottle |
CAS Min % | 88.0 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Formula Weight | 160.17 |
Physical Form | Liquid |
Show More |
Safety and Handling
GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.