Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethoxyaniline, 99%, Acros Organics™

Click to view available options
Quantity:
5g
50g
250g
Packaging:
Glass bottle
102-56-7
C8H11NO2
153.18
MFCD00008368
NAZDVUBIEPVUKE-UHFFFAOYSA-N
benzenamine, 2,5-dimethoxy,aminohydroquinone dimethyl ether,1-amino-2,5-dimethoxybenzene,2,5-dimethoxybenzenamine,aniline, 2,5-dimethoxy,unii-v3z5u3fl10,2,5 dimethoxyaniline,2,5-dimethoxyphenylamine,dimethoxyaniline 2,5-,2,5-dimethoxy aniline
7613
2,5-dimethoxyaniline
COC1=CC(=C(C=C1)OC)N
102-56-7
C8H11NO2
153.18
MFCD00008368
NAZDVUBIEPVUKE-UHFFFAOYSA-N
benzenamine, 2,5-dimethoxy,aminohydroquinone dimethyl ether,1-amino-2,5-dimethoxybenzene,2,5-dimethoxybenzenamine,aniline, 2,5-dimethoxy,unii-v3z5u3fl10,2,5 dimethoxyaniline,2,5-dimethoxyphenylamine,dimethoxyaniline 2,5-,2,5-dimethoxy aniline
7613
2,5-dimethoxyaniline
COC1=CC(=C(C=C1)OC)N
Boiling Point 140.0°C (7.0 mmHg)
Melting Point 79.0°C to 82.0°C
Flash Point 130°C
Quantity 250g
Packaging Glass bottle
Color Brown to Gray
Infrared Spectrum Authentic
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3O)2C6H3NH2
Beilstein 13, 788
Solubility Information Solubility in water: 4g/L (40°C). Other solubilities: soluble in most common organic solvents
Formula Weight 153.18
Physical Form Crystalline Powder or Lumps
Percent Purity 99%
Chemical Name or Material 2, 5-Dimethoxyaniline, 99%
Show More Show Less

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 203-040-9

RTECSNumber : BX4375000

TSCA : TSCA