Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethoxybenzaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
93-02-7
C9H10O3
166.18
AFUKNJHPZAVHGQ-UHFFFAOYSA-N
benzaldehyde, 2,5-dimethoxy,2,5-dimethoxy benzaldehyde,unii-w49s1ppl78,2,5-dimethoxybenzald,pubchem2176,gentisic aldehyde,5-dimethoxy benzaldehyde,2,5-dimethoxybenzaldehyd,2,5-dimethoxybenzaldehye,2.5-dimethoxybenzaldehyde
66726
2,5-dimethoxybenzaldehyde
COC1=CC(=C(C=C1)OC)C=O
93-02-7
C9H10O3
166.18
AFUKNJHPZAVHGQ-UHFFFAOYSA-N
benzaldehyde, 2,5-dimethoxy,2,5-dimethoxy benzaldehyde,unii-w49s1ppl78,2,5-dimethoxybenzald,pubchem2176,gentisic aldehyde,5-dimethoxy benzaldehyde,2,5-dimethoxybenzaldehyd,2,5-dimethoxybenzaldehye,2.5-dimethoxybenzaldehyde
66726
2,5-dimethoxybenzaldehyde
COC1=CC(=C(C=C1)OC)C=O
Melting Point 45.0°C to 50.0°C
Quantity 100g
Boiling Point 146.0°C (10.0 mmHg)
Color Yellow to Beige
Flash Point 113°C
Infrared Spectrum Authentic
Packaging Glass bottle
Water 0.5% max.
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CH3O)2C6H3CHO
Beilstein 08,245
Solubility Information Solubility in water: 795mg/l (25°C). Other solubilities: soluble in most common organic solvents
Formula Weight 166.18
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethoxybenzaldehyde
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning