Alfa Aesar™ 2,5-Dimethoxybenzenesulfonamide, 97%

Manufacturer: Alfa Aesar™ B2264903

Quantity 1g

 View more versions of this product

Catalog No. AAB2264903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19116-90-6
    Molecular Formula C8H11NO4S
    Formula Weight 217.25
    MDL Number MFCD00833415
    Chemical Name or Material 2,5-Dimethoxybenzenesulfonamide

    Specifications

    Formula Weight 217.25
    Molecular Formula C8H11NO4S
    Melting Point 145° to 149°C
    MDL Number MFCD00833415
    Quantity 1g
    RTECS Number DB2030000
    CAS 19116-90-6
    Chemical Name or Material 2,5-Dimethoxybenzenesulfonamide
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit