Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethoxybenzoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2785-98-0
C9H10O4
182.175
MFCD00002436
NYJBTJMNTNCTCP-UHFFFAOYSA-N
benzoic acid, 2,5-dimethoxy,6-methoxy-m-anisic acid,2,5-dimethoxy-benzoic acid,2,5-dimethoxybenzoicacid,pubchem14413,acmc-1cit0,2,5-dimethoxybenzoic acid
76027
2,5-dimethoxybenzoic acid
COC1=CC(=C(C=C1)OC)C(=O)O
2785-98-0
C9H10O4
182.175
MFCD00002436
NYJBTJMNTNCTCP-UHFFFAOYSA-N
benzoic acid, 2,5-dimethoxy,6-methoxy-m-anisic acid,2,5-dimethoxy-benzoic acid,2,5-dimethoxybenzoicacid,pubchem14413,acmc-1cit0,2,5-dimethoxybenzoic acid
76027
2,5-dimethoxybenzoic acid
COC1=CC(=C(C=C1)OC)C(=O)O
Quantity 5g
Melting Point 74°C to 78°C
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3O)2C6H3COOH
Formula Weight 182.18
Physical Form Fine Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2, 5-Dimethoxybenzoic acid, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 220-503-0

TSCA : TSCA