Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Dimethoxyphenyl isothiocyanate, 98%

Click to view available options
Quantity:
10g
50g
40532-06-7
C9H9NO2S
195.236
MFCD00041059
RADQLZMDTYWZKA-UHFFFAOYSA-N
2,5-dimethoxyphenyl isothiocyanate,2,5-dimethoxyphenylisothiocyanate,2-isothiocyanato-1,4-dimethoxy-benzene,benzene, 2-isothiocyanato-1,4-dimethoxy,2,5-dimethoxybenzenisothiocyanate,acmc-1amrr,#
142466
2-isothiocyanato-1,4-dimethoxybenzene
COC1=CC(=C(C=C1)OC)N=C=S
40532-06-7
C9H9NO2S
195.236
MFCD00041059
RADQLZMDTYWZKA-UHFFFAOYSA-N
2,5-dimethoxyphenyl isothiocyanate,2,5-dimethoxyphenylisothiocyanate,2-isothiocyanato-1,4-dimethoxy-benzene,benzene, 2-isothiocyanato-1,4-dimethoxy,2,5-dimethoxybenzenisothiocyanate,acmc-1amrr,#
142466
2-isothiocyanato-1,4-dimethoxybenzene
COC1=CC(=C(C=C1)OC)N=C=S
Boiling Point 174°C to 175°C (10mmHg)
Sensitivity Moisture sensitive
Flash Point >110°C (230°F)
Melting Point 35°C to 37°C
Quantity 10g
UN Number UN2923
Beilstein 3267310
Formula Weight 195.24
Percent Purity 98%
Chemical Name or Material 2,5-Dimethoxyphenyl isothiocyanate

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures