Learn More
eMolecules​ 2,5-DIMETHYL-1,4-BENZOQUINONE | 137-18-8 | 1G | Purity: 98% DFS Item
Manufacturer:  eMolecules​ 384829566
AstaTech | 2,5-DIMETHYL-1,4-BENZOQUINONE | 1G | 137-18-8 | MFCD00041737
Catalog No. 50-158-0839
Due to product restrictions, please to purchase or view availability for this product.