Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Dimethyl-1-(4-nitrophenyl)pyrrole, 97%

Click to view available options
Quantity:
1g
5g
5044-22-4
C12H12N2O2
216.24
MFCD00085038
JXJRCFOGAIXRCU-UHFFFAOYSA-N
2,5-dimethyl-1-4-nitrophenyl-1h-pyrrole,2,5-dimethyl-1-4-nitrophenyl pyrrole,1-4-nitrophenyl-2,5-dimethylpyrrole,maybridge1_007286,acmc-1auq2,2,5-dimethyl-1-p-nitrophenyl pyrrole,n-4'-nitrophenyl-2,5-dimethylpyrrole,1-4-nitrophenyl-2,5-dimethyl-1h-pyrrole
459085
2,5-dimethyl-1-(4-nitrophenyl)pyrrole
CC1=CC=C(N1C2=CC=C(C=C2)[N+](=O)[O-])C
5044-22-4
C12H12N2O2
216.24
MFCD00085038
JXJRCFOGAIXRCU-UHFFFAOYSA-N
2,5-dimethyl-1-4-nitrophenyl-1h-pyrrole,2,5-dimethyl-1-4-nitrophenyl pyrrole,1-4-nitrophenyl-2,5-dimethylpyrrole,maybridge1_007286,acmc-1auq2,2,5-dimethyl-1-p-nitrophenyl pyrrole,n-4'-nitrophenyl-2,5-dimethylpyrrole,1-4-nitrophenyl-2,5-dimethyl-1h-pyrrole
459085
2,5-dimethyl-1-(4-nitrophenyl)pyrrole
CC1=CC=C(N1C2=CC=C(C=C2)[N+](=O)[O-])C
Quantity 1g
Melting Point 145°C to 148°C
Formula Weight 216.24
Percent Purity 97%
Chemical Name or Material 2,5-Dimethyl-1-(4-nitrophenyl)pyrrole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning