Alfa Aesar™ 2,5-Dimethyl-1,5-hexadiene, 98%

Manufacturer: Alfa Aesar™ B2178118

Quantity 50g

 View more versions of this product

Catalog No. AAB2178118

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 627-58-7
    Molecular Formula C8H14
    Formula Weight 110.2
    MDL Number MFCD00008602
    UN Number UN3295
    Beilstein 1697948
    Chemical Name or Material 2,5-Dimethyl-1,5-hexadiene

    Specifications

    Formula Weight 110.2
    TSCA TSCA
    Molecular Formula C8H14
    MDL Number MFCD00008602
    UN Number UN3295
    Beilstein 1697948
    Health Hazard 2 GHS H Statement
    H225-H304-H315-H319-H335
    Highly flammable liquid and vapour.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
    Flash Point 13°C (55°F)
    Odor Characteristic
    Boiling Point 114° to 116°C
    Melting Point −75°C
    Quantity 50g
    CAS 627-58-7
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2,5-Dimethyl-1,5-hexadiene
    Assay 98%

    GHS H Statement
    H225-H304-H315-H319-H335
    Highly flammable liquid and vapour.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit