Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane, 92%, Acros Organics

Click to view available options
Quantity:
5g
100g
Packaging:
Plastic bottle
78-63-7
C16H34O4
290.44
DMWVYCCGCQPJEA-UHFFFAOYSA-N
varox,2,5-dimethyl-2,5-di tert-butylperoxy hexane,2,5-bis tert-butylperoxy-2,5-dimethylhexane,triqanox xq 8,kayahexa ad 40c,lupersol 101,luperco 101xl,luperox 101,perhexa 3m40,varox 50
6545
2,5-bis(tert-butylperoxy)-2,5-dimethylhexane
CC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C
78-63-7
C16H34O4
290.44
DMWVYCCGCQPJEA-UHFFFAOYSA-N
varox,2,5-dimethyl-2,5-di tert-butylperoxy hexane,2,5-bis tert-butylperoxy-2,5-dimethylhexane,triqanox xq 8,kayahexa ad 40c,lupersol 101,luperco 101xl,luperox 101,perhexa 3m40,varox 50
6545
2,5-bis(tert-butylperoxy)-2,5-dimethylhexane
CC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C
Radical initiators
Specific Gravity 0.87
Viscosity 6.4 mPa.s (20°C)
Quantity 100g
Density 0.8700g/mL
Refractive Index 1.4200 to 1.4240
Packaging Plastic bottle
Flash Point 68°C
Boiling Point 55.0°C to 57.0°C (7.0mmHg)
Melting Point 6.0°C
Assay Percent Range 92%
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in organic solvents,soluble in alcohol
Formula Weight 290.44
Percent Purity 92%
Chemical Name or Material 2, 5-Dimethyl-2, 5-di(tert-butylperoxy)hexane
Show More Show Less