Alfa Aesar™ 2,5-Dimethyl-2,5-hexanediol, 97%

Manufacturer: Alfa Aesar™ A119840E

Quantity 2500g

 View more versions of this product

Catalog No. AAA119840E

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 110-03-2
    Molecular Formula C8H18O2
    Formula Weight 146.23
    MDL Number MFCD00004473
    Beilstein 1361437
    Chemical Name or Material 2,5-Dimethyl-2,5-hexanediol

    Specifications

    Formula Weight 146.23
    Boiling Point 214° to 215°C
    Quantity 2500g
    Flash Point 126°C (258°F)
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P305+P351+P338
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    TSCA TSCA
    Molecular Formula C8H18O2
    Melting Point 87° to 91°C
    MDL Number MFCD00004473
    CAS 110-03-2
    Beilstein 1361437
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    Chemical Name or Material 2,5-Dimethyl-2,5-hexanediol
    Assay 97%

    Description

    Applications
    It is used as an intermediate in the preparation of 2.5-Dimethyl-2.5-bis(tertbutyl-peroxy)hexane to produce polyethylene copolymers and polyethylene rubbers. It is also used in the synthesis of six- and seven-membered heterocyclic boron compounds containing intramolecular N-B bond.

    Solubility
    Soluble in water 20°C 140g/L.

    Notes
    Store away from acid chloride, acid anhydrides and oxidizing agents. Keep the container tightly closed.

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P305+P351+P338
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit