Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethyl-3-hexyne-2,5-diol, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
142-30-3
C8H14O2
142.2
MFCD00004468
IHJUECRFYCQBMW-UHFFFAOYSA-N
2,5-dimethyl-3-hexyne-2,5-diol, dimethylhexynediol, kemitracin-50, tetramethylbutynediol, acetylenepinacol, 3-hexyne-2,5-diol, 2,5-dimethyl, olfine y, tetramethyl-2-butynediol, unii-22rr53u71w, 2,5-dimethylhexyne-2,5-diol
8883
2,5-dimethylhex-3-yne-2,5-diol
CC(C)(C#CC(C)(C)O)O
142-30-3
C8H14O2
142.2
MFCD00004468
IHJUECRFYCQBMW-UHFFFAOYSA-N
2,5-dimethyl-3-hexyne-2,5-diol, dimethylhexynediol, kemitracin-50, tetramethylbutynediol, acetylenepinacol, 3-hexyne-2,5-diol, 2,5-dimethyl, olfine y, tetramethyl-2-butynediol, unii-22rr53u71w, 2,5-dimethylhexyne-2,5-diol
8883
2,5-dimethylhex-3-yne-2,5-diol
CC(C)(C#CC(C)(C)O)O
Melting Point >89.0°C
Boiling Point 121.0°C to 123.0°C (7.0 mmHg)
Color White to Yellow
Packaging Plastic Bottle
Infrared Spectrum Authentic
Quantity 500g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula HOC(CH3)2C≡CC(CH3)2OH
Beilstein 01, 501
Solubility Information Solubility in water: 240g/L (20 c). Other solubilities: soluble in acetone,alcohol and ethyl acetate,slightly soluble in benzene,naphtha and,carbon tetrachloride
Formula Weight 142.2
Physical Form Crystalline Flakes or Powder
Percent Purity 98%
Chemical Name or Material 2, 5-Dimethyl-3-hexyne-2, 5-diol
Show More Show Less

EINECSNumber : 205-533-4