Alfa Aesar™ 2',5'-Dimethylacetophenone, 98+%

Manufacturer: Alfa Aesar™ A1143214

Quantity 25g

 View more versions of this product

Catalog No. AAA1143214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2142-73-6
    Molecular Formula C10H12O
    Formula Weight 148.21
    MDL Number MFCD00014988
    Beilstein 2042139
    Chemical Name or Material 2',5'-Dimethylacetophenone

    Specifications

    Formula Weight 148.21
    CAS 2142-73-6
    Molecular Formula C10H12O
    Boiling Point 230° to 231°C
    Beilstein 2042139
    Quantity 25g
    Flash Point 95°C (203°F)
    TSCA TSCA
    MDL Number MFCD00014988
    Odor Characteristic
    Chemical Name or Material 2',5'-Dimethylacetophenone
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit