Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylaniline 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
95-78-3
C8H11N
121.18
MFCD00007743
VOWZNBNDMFLQGM-UHFFFAOYSA-N
2,5-xylidine,p-xylidine,2,5-dimethylphenylamine,2-amino-1,4-xylene,2,5-dimethylbenzenamine,benzenamine, 2,5-dimethyl,5-methyl-o-toluidine,6-methyl-m-toluidine,1-amino-2,5-dimethylbenzene,p-dimethylaniline
7259
CHEBI:518305
2,5-dimethylaniline
CC1=CC(=C(C=C1)C)N
95-78-3
C8H11N
121.18
MFCD00007743
VOWZNBNDMFLQGM-UHFFFAOYSA-N
2,5-xylidine,p-xylidine,2,5-dimethylphenylamine,2-amino-1,4-xylene,2,5-dimethylbenzenamine,benzenamine, 2,5-dimethyl,5-methyl-o-toluidine,6-methyl-m-toluidine,1-amino-2,5-dimethylbenzene,p-dimethylaniline
7259
CHEBI:518305
2,5-dimethylaniline
CC1=CC(=C(C=C1)C)N
Boiling Point 218°C
Melting Point 11.5°C
Flash Point 93°C
Specific Gravity 0.97
Refractive Index 1.5582 to 1.5602
Density 0.9700g/mL
Packaging Glass bottle
Quantity 500g
Color Undesignated
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2C6H3NH2
Beilstein 12, 1135
Merck Index 15, 10282
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,carbon tetrachloride
Formula Weight 121.18
Physical Form Liquid
Percent Purity 98.5 to 101.5% total base, ≥98.5% (GC)
Chemical Name or Material 2, 5-Dimethylaniline, 99%
Show More Show Less

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-451-0

RTECSNumber : ZE9100000

TSCA : TSCA

Recommended Storage : product darkens if exposed to light and air