Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Dimethylbenzoxazole, 99%

Click to view available options
Quantity:
1g
5g
5676-58-4
C9H9NO
147.177
MFCD00005773
XVQGFGKAPKEUFT-UHFFFAOYSA-N
2,5-dimethylbenzoxazole,2,5-dimethylbenzo d oxazole,benzoxazole, 2,5-dimethyl,2,5-dimethyl benzoxazole,pubchem8640,acmc-209lua,benzoxazole,5-dimethyl,2,5-dimethyl-benzoxazole,benzoxazole,2,5-dimethyl,#
21884
2,5-dimethyl-1,3-benzoxazole
CC1=CC2=C(C=C1)OC(=N2)C
5676-58-4
C9H9NO
147.177
MFCD00005773
XVQGFGKAPKEUFT-UHFFFAOYSA-N
2,5-dimethylbenzoxazole,2,5-dimethylbenzo d oxazole,benzoxazole, 2,5-dimethyl,2,5-dimethyl benzoxazole,pubchem8640,acmc-209lua,benzoxazole,5-dimethyl,2,5-dimethyl-benzoxazole,benzoxazole,2,5-dimethyl,#
21884
2,5-dimethyl-1,3-benzoxazole
CC1=CC2=C(C=C1)OC(=N2)C
Boiling Point 218°C to 219°C
Quantity 5g
Formula Weight 147.18
Percent Purity 99%
Chemical Name or Material 2,5-Dimethylbenzoxazole

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA