Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylbenzoxazole 99%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
5676-58-4
C9H9NO
147.177
MFCD00005773
XVQGFGKAPKEUFT-UHFFFAOYSA-N
2,5-dimethylbenzoxazole,2,5-dimethylbenzo d oxazole,benzoxazole, 2,5-dimethyl,2,5-dimethyl benzoxazole,pubchem8640,acmc-209lua,benzoxazole,5-dimethyl,2,5-dimethyl-benzoxazole,benzoxazole,2,5-dimethyl,#
21884
2,5-dimethyl-1,3-benzoxazole
CC1=CC2=C(C=C1)OC(=N2)C
5676-58-4
C9H9NO
147.177
MFCD00005773
XVQGFGKAPKEUFT-UHFFFAOYSA-N
2,5-dimethylbenzoxazole,2,5-dimethylbenzo d oxazole,benzoxazole, 2,5-dimethyl,2,5-dimethyl benzoxazole,pubchem8640,acmc-209lua,benzoxazole,5-dimethyl,2,5-dimethyl-benzoxazole,benzoxazole,2,5-dimethyl,#
21884
2,5-dimethyl-1,3-benzoxazole
CC1=CC2=C(C=C1)OC(=N2)C
Density 1.0900g/mL
Refractive Index 1.5425 to 1.5445 (20°C, 589nm)
Quantity 10g
Color Yellow to Brown
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 27, 49
Formula Weight 147.18
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2, 5-Dimethylbenzoxazole, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 227-136-5

RTECSNumber : DM4821600

TSCA : TSCA