Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylfuran, 99%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass Bottle
625-86-5
C6H8O
96.13
MFCD00003250
GSNUFIFRDBKVIE-UHFFFAOYSA-N
furan, 2,5-dimethyl,2,5-dimethylfurane,2,5-dimethyl-furan,2,5-dimethyl furane,unii-dr5hl9oj7y,ccris 3158,2,5-dimethyl furan,dr5hl9oj7y,2.5-dimethyl furan,acmc-1avpg
12266
2,5-dimethylfuran
CC1=CC=C(O1)C
625-86-5
C6H8O
96.13
MFCD00003250
GSNUFIFRDBKVIE-UHFFFAOYSA-N
furan, 2,5-dimethyl,2,5-dimethylfurane,2,5-dimethyl-furan,2,5-dimethyl furane,unii-dr5hl9oj7y,ccris 3158,2,5-dimethyl furan,dr5hl9oj7y,2.5-dimethyl furan,acmc-1avpg
12266
2,5-dimethylfuran
CC1=CC=C(O1)C
Density 0.9000g/mL
Melting Point -62.0°C
Specific Gravity 0.9
Color Colorless to Amber
Quantity 500mL
Packaging Glass Bottle
Refractive Index 1.4400 to 1.4420
Infrared Spectrum Authentic
Flash Point −1°C
Boiling Point 92.0°C to 94.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
UN Number 1992
Beilstein 17,41
Merck Index 15,3268
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in oil,organic solvents
Formula Weight 96.13
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2, 5-Dimethylfuran, 99%
Show More Show Less

GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Highly flammable liquid and vapour.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 210-914-3

RTECSNumber : LU0875000

TSCA : TSCA

Recommended Storage : May darken on storage