Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylindole, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1196-79-8
C10H11N
145.2
MFCD00005621
ZFLFWZRPMDXJCW-UHFFFAOYSA-N
2,5-dimethylindole,1h-indole, 2,5-dimethyl,indole, 2,5-dimethyl,unii-etu94v283m,2,5-dimethyl indole,pubchem7330,acmc-1bvpm
70965
2,5-dimethyl-1H-indole
CC1=CC2=C(C=C1)NC(=C2)C
1196-79-8
C10H11N
145.2
MFCD00005621
ZFLFWZRPMDXJCW-UHFFFAOYSA-N
2,5-dimethylindole,1h-indole, 2,5-dimethyl,indole, 2,5-dimethyl,unii-etu94v283m,2,5-dimethyl indole,pubchem7330,acmc-1bvpm
70965
2,5-dimethyl-1H-indole
CC1=CC2=C(C=C1)NC(=C2)C
Infrared Spectrum Authentic
Melting Point 112.0°C to 113.0°C
Quantity 1g
Packaging Glass bottle
Color Brown or Pink to White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 20, 320
Formula Weight 145.2
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethylindole
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.