Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylphenylacetic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
13612-34-5
C10H12O2
164.204
MFCD00014369
RUSCTNYOPQOXDJ-UHFFFAOYSA-N
2,5-dimethylphenylacetic acid,2-2,5-dimethylphenyl acetic acid,2,5-dimethylphenyl acetic acid,2,5-xylylacetic acid,2,5-dimethylphenylaceticacid,2,5-dimethylbenzeneacetic acid,2,5-dimethylphenyl acetic acid,benzeneacetic acid, 2,5-dimethyl,pubchem22525,acmc-1bb4h
83615
2-(2,5-dimethylphenyl)acetic acid
CC1=CC(=C(C=C1)C)CC(=O)O
13612-34-5
C10H12O2
164.204
MFCD00014369
RUSCTNYOPQOXDJ-UHFFFAOYSA-N
2,5-dimethylphenylacetic acid,2-2,5-dimethylphenyl acetic acid,2,5-dimethylphenyl acetic acid,2,5-xylylacetic acid,2,5-dimethylphenylaceticacid,2,5-dimethylbenzeneacetic acid,2,5-dimethylphenyl acetic acid,benzeneacetic acid, 2,5-dimethyl,pubchem22525,acmc-1bb4h
83615
2-(2,5-dimethylphenyl)acetic acid
CC1=CC(=C(C=C1)C)CC(=O)O
Packaging Glass bottle
Melting Point 128°C to 132°C
Quantity 1g
Color Beige
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 09, IV, 1850
Formula Weight 164.2
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2, 5-Dimethylphenylacetic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.