Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
85199-06-0
C8H11BO2
149.98
MFCD01863525
OOMZKLJLVGQZGV-UHFFFAOYSA-N
2,5-dimethylphenyl boronic acid,2,5-dimethylbenzeneboronic acid,2,5-dimethylphenylboronicacid,2,5-dimethylphenyl boranediol,p-xylene-2-boronic acid,boronic acid, 2,5-dimethylphenyl,pubchem1831,p-xyleneboronic acid,ksc448a1r,2,5-dimehylphenylboronic acid
2734347
(2,5-dimethylphenyl)boronic acid
B(C1=C(C=CC(=C1)C)C)(O)O
85199-06-0
C8H11BO2
149.98
MFCD01863525
OOMZKLJLVGQZGV-UHFFFAOYSA-N
2,5-dimethylphenyl boronic acid,2,5-dimethylbenzeneboronic acid,2,5-dimethylphenylboronicacid,2,5-dimethylphenyl boranediol,p-xylene-2-boronic acid,boronic acid, 2,5-dimethylphenyl,pubchem1831,p-xyleneboronic acid,ksc448a1r,2,5-dimehylphenylboronic acid
2734347
(2,5-dimethylphenyl)boronic acid
B(C1=C(C=CC(=C1)C)C)(O)O
Quantity 5g
Melting Point 186.0°C to 191.0°C
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 149.98
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethylphenylboronic acid

GHS P Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.