Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylphenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
85199-06-0
C8H11BO2
149.98
MFCD01863525
OOMZKLJLVGQZGV-UHFFFAOYSA-N
2,5-dimethylphenyl boronic acid, 2,5-dimethylbenzeneboronic acid, 2,5-dimethylphenylboronicacid, 2,5-dimethylphenyl boranediol, p-xylene-2-boronic acid, boronic acid, 2,5-dimethylphenyl, pubchem1831, p-xyleneboronic acid, ksc448a1r, 2,5-dimehylphenylboronic acid
2734347
(2,5-dimethylphenyl)boronic acid
CC1=CC=C(C)C(=C1)B(O)O
85199-06-0
C8H11BO2
149.98
MFCD01863525
OOMZKLJLVGQZGV-UHFFFAOYSA-N
2,5-dimethylphenyl boronic acid, 2,5-dimethylbenzeneboronic acid, 2,5-dimethylphenylboronicacid, 2,5-dimethylphenyl boranediol, p-xylene-2-boronic acid, boronic acid, 2,5-dimethylphenyl, pubchem1831, p-xyleneboronic acid, ksc448a1r, 2,5-dimehylphenylboronic acid
2734347
(2,5-dimethylphenyl)boronic acid
CC1=CC=C(C)C(=C1)B(O)O
Melting Point 186.0°C to 191.0°C
Packaging Glass bottle
Quantity 25g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 149.98
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethylphenylboronic acid

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning