Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylphenylhydrazine Hydrochloride, ACROS Organics™

Click to view available options
Quantity:
5g
25g
56737-78-1
C8H13ClN2
172.656
MFCD00013382
OMOFMSZLYCEIAF-UHFFFAOYSA-N
2,5-dimethylphenylhydrazine hydrochloride,2,5-dimethylphenyl hydrazine hydrochloride,2,5-dimethylphenylhydrazinehydrochloride,2,3-dimethylphenylhydrazinehydrochloride,pubchem6400,acmc-1bf0t,ksc491e8b,2-hydrazino-m-xylene hydrochloride,2,5-dimethyl-phenylhydrazine hydrochloride,1-2,5-dimethylphenyl hydrazine hydrochloride
2734058
(2,5-dimethylphenyl)hydrazine;hydrochloride
CC1=CC(=C(C=C1)C)NN.Cl
56737-78-1
C8H13ClN2
172.656
MFCD00013382
OMOFMSZLYCEIAF-UHFFFAOYSA-N
2,5-dimethylphenylhydrazine hydrochloride,2,5-dimethylphenyl hydrazine hydrochloride,2,5-dimethylphenylhydrazinehydrochloride,2,3-dimethylphenylhydrazinehydrochloride,pubchem6400,acmc-1bf0t,ksc491e8b,2-hydrazino-m-xylene hydrochloride,2,5-dimethyl-phenylhydrazine hydrochloride,1-2,5-dimethylphenyl hydrazine hydrochloride
2734058
(2,5-dimethylphenyl)hydrazine;hydrochloride
CC1=CC(=C(C=C1)C)NN.Cl
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Formula Weight 172.65
Chemical Name or Material 2, 5-Dimethylphenylhydrazine hydrochloride