Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2,5-Dimethylphenylhydrazine Hydrochloride, ACROS Organics™
Click to view available options
Quantity:
5g
25g
Specifications
Quantity | 25g |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Formula Weight | 172.65 |
Chemical Name or Material | 2, 5-Dimethylphenylhydrazine hydrochloride |
Safety and Handling
TSCA : TSCA