Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylpiperazine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
2815-34-1
C6H14N2
114.192
NSMWYRLQHIXVAP-OLQVQODUSA-N
trans-2,5-dimethylpiperazine,2r,5s-2,5-dimethylpiperazine,2s,5r-2,5-dimethylpiperazine,2s,5r-2,5-dimethyl-piperazine,piperazine, 2,5-dimethyl-, 2r,5s-rel,2r,5s-2,5-dimethylpiperazin,piperazine, 2,5-dimethyl-, trans,2r,5s-2,5-dimethyl-piperazine,pubchem15878,2,5-dimethylpiperazine #
220672
(2R,5S)-2,5-dimethylpiperazine
CC1CNC(CN1)C
2815-34-1
C6H14N2
114.192
NSMWYRLQHIXVAP-OLQVQODUSA-N
trans-2,5-dimethylpiperazine,2r,5s-2,5-dimethylpiperazine,2s,5r-2,5-dimethylpiperazine,2s,5r-2,5-dimethyl-piperazine,piperazine, 2,5-dimethyl-, 2r,5s-rel,2r,5s-2,5-dimethylpiperazin,piperazine, 2,5-dimethyl-, trans,2r,5s-2,5-dimethyl-piperazine,pubchem15878,2,5-dimethylpiperazine #
220672
(2R,5S)-2,5-dimethylpiperazine
CC1CNC(CN1)C
Quantity 5g
Melting Point 113°C to 118°C
Infrared Spectrum Authentic
Density 0.5300g/mL
Color White to Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 23, 19
Formula Weight 114.19
Physical Form Crystalline Powder or Crystals
Percent Purity ≥97.5%
Chemical Name or Material trans-2, 5-Dimethylpiperazine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Flammable solid.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger