Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylpyrazine, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
123-32-0
C6H8N2
108.14
MFCD00006147
LCZUOKDVTBMCMX-UHFFFAOYSA-N
2,5-dimethyl pyrazine,pyrazine, 2,5-dimethyl,2,5-dimethylpiazine,2,5-dimethyl-1,4-diazine,2,5-dimethylparadiazine,2,5-dimethyl-pyrazine,unii-v99y0muy1q,fema no. 3272,ccris 2929,natural
31252
2,5-dimethylpyrazine
CC1=CN=C(C=N1)C
123-32-0
C6H8N2
108.14
MFCD00006147
LCZUOKDVTBMCMX-UHFFFAOYSA-N
2,5-dimethyl pyrazine,pyrazine, 2,5-dimethyl,2,5-dimethylpiazine,2,5-dimethyl-1,4-diazine,2,5-dimethylparadiazine,2,5-dimethyl-pyrazine,unii-v99y0muy1q,fema no. 3272,ccris 2929,natural
31252
2,5-dimethylpyrazine
CC1=CN=C(C=N1)C
Density 0.9900g/mL
Refractive Index 1.4987 to 1.5007
Flash Point 63°C
Boiling Point 155°C
Specific Gravity 0.99
Infrared Spectrum Authentic
Color Colorless to Yellow
Packaging Glass bottle
Quantity 25g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 23, 96
Solubility Information Solubility in water: soluble. Other solubilities: soluble in organic solvents,miscible with ethanol,ethylether,acetone
Formula Weight 108.14
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2, 5-Dimethylpyrazine, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/pro

GHS Signal Word: Warning

EINECSNumber : 204-618-3

RTECSNumber : UQ2800000

TSCA : TSCA