Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylpyrrole, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
625-84-3
C6H9N
95.145
MFCD00005223
PAPNRQCYSFBWDI-UHFFFAOYSA-N
2,5-dimethylpyrrole,2,5-dimethylazole,1h-pyrrole, 2,5-dimethyl,pyrrole, 2,5-dimethyl,unii-mz3oyf5521,2,5-dimethyl-pyrrole,pyrrole,2,5-dimethyl,2,5-dimethyl pyrrole,pyrrole, 2,5-dimethyl-8ci,2,5-bimethylpyrrole
12265
2,5-dimethyl-1H-pyrrole
CC1=CC=C(N1)C
625-84-3
C6H9N
95.145
MFCD00005223
PAPNRQCYSFBWDI-UHFFFAOYSA-N
2,5-dimethylpyrrole,2,5-dimethylazole,1h-pyrrole, 2,5-dimethyl,pyrrole, 2,5-dimethyl,unii-mz3oyf5521,2,5-dimethyl-pyrrole,pyrrole,2,5-dimethyl,2,5-dimethyl pyrrole,pyrrole, 2,5-dimethyl-8ci,2,5-bimethylpyrrole
12265
2,5-dimethyl-1H-pyrrole
CC1=CC=C(N1)C
Refractive Index 1.5040 to 1.5080 (20°C, 589nm)
Density 0.935g/mL
Quantity 1g
Infrared Spectrum Authentic
Color Orange-Yellow to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 20, 172
Formula Weight 95.14
Physical Form Crystalline Powder and/or Chunks
Percent Purity ≥96.0% (GC)
Chemical Name or Material 2, 5-Dimethylpyrrole
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Recommended Storage : Product may darken during storage