Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylpyrrole, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
625-84-3
C6H9N
95.14
MFCD00005223
PAPNRQCYSFBWDI-UHFFFAOYSA-N
2,5-dimethylpyrrole,2,5-dimethylazole,1h-pyrrole, 2,5-dimethyl,pyrrole, 2,5-dimethyl,unii-mz3oyf5521,2,5-dimethyl-pyrrole,pyrrole,2,5-dimethyl,2,5-dimethyl pyrrole,pyrrole, 2,5-dimethyl-8ci,2,5-bimethylpyrrole
12265
2,5-dimethyl-1H-pyrrole
CC1=CC=C(N1)C
625-84-3
C6H9N
95.14
MFCD00005223
PAPNRQCYSFBWDI-UHFFFAOYSA-N
2,5-dimethylpyrrole,2,5-dimethylazole,1h-pyrrole, 2,5-dimethyl,pyrrole, 2,5-dimethyl,unii-mz3oyf5521,2,5-dimethyl-pyrrole,pyrrole,2,5-dimethyl,2,5-dimethyl pyrrole,pyrrole, 2,5-dimethyl-8ci,2,5-bimethylpyrrole
12265
2,5-dimethyl-1H-pyrrole
CC1=CC=C(N1)C
Specific Gravity 0.935
Quantity 25g
Infrared Spectrum Authentic
Boiling Point 165.0°C (740.0 mmHg)
Color Orange-Yellow to Yellow
Packaging Glass bottle
Flash Point 54°C
Density 0.9350g/mL
Refractive Index 1.5040 to 1.5070
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 20, 172
Solubility Information Solubility in water: insoluble
Formula Weight 95.14
Physical Form Crystalline Powder and/or Chunks
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethylpyrrole
Show More Show Less

GHS H StatementToxic if inhaled.Toxic if swallowed.Toxic in contact with skin.Causes skin irritation.May cause respiratory irritation.Causes serious eye irritation.Flammable liquid and vapor.

GHS P StatementKeep away from heat/sparks/open flames/hot surfaces.- No smoking.IF ON SKIN (or hair): Take off immediately all contaminated clothing.Rinse skin with water/shower.Wash face,hands and any exposed skin thoroughly after handling.IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.Call a POISON CENTER or doctor/physician.Wear protective gloves/protective clothing/eye protection/face protection.If skin irritation occurs: Get medical advice/attention.If eye irritation persists: Get medical advice/attention.

;GHS P StatementKeep away from heat/sparks/open flames/hot surfaces.- No smoking.IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.Wash face,hands and any exposed skin thoroughly after handli

Recommended Storage : Product may darken during storage