Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylpyrrolidine, 97%, Maybridge

Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
3378-71-0
C6H13N
99.177
MFCD00005254
ZEBFPAXSQXIPNF-UHFFFAOYSA-N
2,5-dimethyl-pyrrolidine,pyrrolidine, 2,5-dimethyl,2rs,5rs-2,5-dimethylpyrrolidine,2,5-dimethylpyrrolidine, mixture of cis and trans, technical grade
96034
2,5-dimethylpyrrolidine
CC1CCC(N1)C
3378-71-0
C6H13N
99.177
MFCD00005254
ZEBFPAXSQXIPNF-UHFFFAOYSA-N
2,5-dimethyl-pyrrolidine,pyrrolidine, 2,5-dimethyl,2rs,5rs-2,5-dimethylpyrrolidine,2,5-dimethylpyrrolidine, mixture of cis and trans, technical grade
96034
2,5-dimethylpyrrolidine
CC1CCC(N1)C
Packaging Amber Glass Bottle
Density 0.81g/mL
Infrared Spectrum Authentic
Color Yellow
Quantity 1g
Refractive Index 1.4290 to 1.4320 (20°C, 589nm)
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 20, 102
Fieser 16,147
Formula Weight 99.18
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2, 5-Dimethylpyrrolidine
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger