Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylthiophene 98.5%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
Packaging:
Glass bottle
638-02-8
C6H8S
112.19
MFCD00005452
GWQOOADXMVQEFT-UHFFFAOYSA-N
thiophene, 2,5-dimethyl,2,5-dimethyl-thiophene,unii-v6ddx6wb12,v6ddx6wb12,pubchem5194,2,5 dimethylthiophene,thiophene,5-dimethyl,2,5-dimethyl thiophene,thiophene,2,5-dimethyl,2,5-dimethyl-2h-thiolium
12514
2,5-dimethylthiophene
CC1=CC=C(S1)C
638-02-8
C6H8S
112.19
MFCD00005452
GWQOOADXMVQEFT-UHFFFAOYSA-N
thiophene, 2,5-dimethyl,2,5-dimethyl-thiophene,unii-v6ddx6wb12,v6ddx6wb12,pubchem5194,2,5 dimethylthiophene,thiophene,5-dimethyl,2,5-dimethyl thiophene,thiophene,2,5-dimethyl,2,5-dimethyl-2h-thiolium
12514
2,5-dimethylthiophene
CC1=CC=C(S1)C
Boiling Point 134°C to 136°C
Flash Point 23°C
Specific Gravity 0.984
Refractive Index 1.5100 to 1.514
Quantity 5mL
Density 0.9840g/mL
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98.5%
Beilstein 17, 41
Solubility Information Solubility in water: insoluble. Other solubilities: in most org solvents
Formula Weight 112.19
Physical Form Liquid
Percent Purity ≥98.0%
Chemical Name or Material 2, 5-Dimethylthiophene, 98.5%
Show More Show Less

GHS P Statement Flammable liquid and vapor.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Warning

EINECSNumber : 211-313-9

TSCA : TSCA