Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,5-Dimethylthiophenol, 98%

Manufacturer:  Alfa Aesar™ B2416406

Quantity 5g

 View more versions of this product

Catalog No. AAB2416406


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

4001-61-0
138.23
UN2810
2,5-Dimethylbenzenethiol
C8H10S
MFCD00010020
2040348
2,5-Dimethylthiophenol

Specifications

4001-61-0
5g
Unpleasant
138.23
UN2810
2,5-Dimethylbenzenethiol
98%
126° to 127°C (50mmHg)
83°C (181°F)
C8H10S
MFCD00010020
2040348
2,5-Dimethylthiophenol

Safety and Handling

SDS & Safety Documents

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit