Alfa Aesar™ 2,5-Dimethylthiophenol, 98%

Manufacturer: Alfa Aesar™ B2416422

Quantity 100g

 View more versions of this product

Catalog No. AAB2416422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4001-61-0
    Molecular Formula C8H10S
    Formula Weight 138.23
    MDL Number MFCD00010020
    UN Number UN2810
    Beilstein 2040348
    Synonym 2,5-Dimethylbenzenethiol
    Chemical Name or Material 2,5-Dimethylthiophenol

    Specifications

    Formula Weight 138.23
    Boiling Point 126° to 127°C (50mmHg)
    Quantity 100g
    Flash Point 83°C (181°F)
    Odor Unpleasant
    Health Hazard 2 GHS H Statement
    H331-H227-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Combustible liquid.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    TSCA TSCA
    Molecular Formula C8H10S
    MDL Number MFCD00010020
    CAS 4001-61-0
    UN Number UN2810
    Beilstein 2040348
    Synonym 2,5-Dimethylbenzenethiol
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P280h-P305+P351+P338-P309-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Chemical Name or Material 2,5-Dimethylthiophenol
    Assay 98%

    GHS H Statement
    H331-H227-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Combustible liquid.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P260-P280h-P305+P351+P338-P309-P310
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit