Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Diphenyl-1,3,4-oxadiazole 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
725-12-2
C14H10N2O
222.247
MFCD00003102
DCJKUXYSYJBBRD-UHFFFAOYSA-N
2,5-diphenyloxadiazole,1,3,4-oxadiazole, 2,5-diphenyl,1,3,4-oxadiazole, 2-5-diphenyl,2,5-diphenyl-1,3,4 oxadiazole,diphenyl-1,3,4-oxadiazole,1,3,4-oxadiazole,2,5-diphenyl,ppd scintillator,acmc-209on9,4-27-00-07212 beilstein handbook reference,wln: t5nn doj cr& er
12888
2,5-diphenyl-1,3,4-oxadiazole
C1=CC=C(C=C1)C2=NN=C(O2)C3=CC=CC=C3
725-12-2
C14H10N2O
222.247
MFCD00003102
DCJKUXYSYJBBRD-UHFFFAOYSA-N
2,5-diphenyloxadiazole,1,3,4-oxadiazole, 2,5-diphenyl,1,3,4-oxadiazole, 2-5-diphenyl,2,5-diphenyl-1,3,4 oxadiazole,diphenyl-1,3,4-oxadiazole,1,3,4-oxadiazole,2,5-diphenyl,ppd scintillator,acmc-209on9,4-27-00-07212 beilstein handbook reference,wln: t5nn doj cr& er
12888
2,5-diphenyl-1,3,4-oxadiazole
C1=CC=C(C=C1)C2=NN=C(O2)C3=CC=CC=C3
Infrared Spectrum Authentic
Quantity 5g
Packaging Glass bottle
Color White
Melting Point 136°C to 141°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 27, 591
Formula Weight 222.25
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 2,5-Diphenyl-1,3,4-oxadiazole, 97%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 211-968-0

RTECSNumber : RO0802500

TSCA : TSCA