Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Diphenyloxazole, Scintillation Grade 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
2.5kg
Packaging:
Plastic bottle
Plastic bucket
92-71-7
C15H11NO
221.259
MFCD00005306
CNRNYORZJGVOSY-UHFFFAOYSA-N
PPO
7105
2,5-diphenyl-1,3-oxazole
C1=CC=C(C=C1)C2=CN=C(O2)C3=CC=CC=C3
92-71-7
C15H11NO
221.259
MFCD00005306
CNRNYORZJGVOSY-UHFFFAOYSA-N
PPO
7105
2,5-diphenyl-1,3-oxazole
C1=CC=C(C=C1)C2=CN=C(O2)C3=CC=CC=C3
for fluorescence/luminescence spectroscopy
Quantity 100g
Additional Information quench parameter to Norm.St.:> 73%
CAS Min % 98.5
Background < 25 cpm
CAS Max % 100.0
Density 1.06g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Beilstein 27, 78
Formula Weight 221.26
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Transmittance 85% min. (40% min. in toluene)
Color White
Melting Point 70°C to 72°C
Chemical Name or Material 2, 5-Diphenyloxazole, 99%
Show More Show Less

EINECSNumber : 202-181-3

RTECSNumber : RP6825000

TSCA : TSCA