Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6,10,14-Tetramethylpentadecane, 95%, ACROS Organics™

Click to view available options
Quantity:
5mL
50mL
100mL
250mL
Packaging:
Glass bottle
1921-70-6
C19H40
268.529
XOJVVFBFDXDTEG-UHFFFAOYSA-N
pristane,norphytane,bute hydrocarbon,norphytan,pristan,pentadecane, 2,6,10,14-tetramethyl,meso-pristane,2,6,10,10-tetramethylpentadecane,meso-form,norphytane, robuoy
15979
CHEBI:53181
2,6,10,14-tetramethylpentadecane
CC(C)CCCC(C)CCCC(C)CCCC(C)C
1921-70-6
C19H40
268.529
XOJVVFBFDXDTEG-UHFFFAOYSA-N
pristane,norphytane,bute hydrocarbon,norphytan,pristan,pentadecane, 2,6,10,14-tetramethyl,meso-pristane,2,6,10,10-tetramethylpentadecane,meso-form,norphytane, robuoy
15979
CHEBI:53181
2,6,10,14-tetramethylpentadecane
CC(C)CCCC(C)CCCC(C)CCCC(C)C
Quantity 100mL
Density 0.7800g/mL
Packaging Glass bottle
Color Colorless
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula (CH3)2CH(CH2)3CH(CH3)(CH2)3CH(CH3)(CH2)3CH(CH3)2
Formula Weight 268.51
Physical Form Liquid
Percent Purity ≥95%
Chemical Name or Material 2,6,10,14-Tetramethylpentadecane, 96%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 217-650-8

RTECSNumber : RZ1880000

TSCA : TSCA