Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Bis[(4R)-phenyl-2-oxazolin-2-yl]pyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
2g
Packaging:
Glass bottle
128249-70-7
C23H19N3O2
369.424
MFCD01863585
HLHBIMJNCKZZQO-SFTDATJTSA-N
2,6-bis 4r-4-phenyl-2-oxazolinyl pyridine,r,r-ph-pybox,2,6-bis r-4-phenyl-4,5-dihydrooxazol-2-yl pyridine,+-2,6-bis 4r-4-phenyl-2-oxazolin-2-yl pyridine,ph-pybox, r,r,2,6-bis 4r-phenyl-2-oxazolin-2-yl pyridine,r,r-2,6-bis 4-phenyl-2-oxazolin-2-yl pyridine,unii-555pn397nx,r,r-2,2'-2,6-pyridinediyl bis 4-phenyl-2-oxazoline,2,6-bis 4r-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl pyridine
2734679
(4R)-4-phenyl-2-[6-[(4R)-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole
C1C(N=C(O1)C2=NC(=CC=C2)C3=NC(CO3)C4=CC=CC=C4)C5=CC=CC=C5
128249-70-7
C23H19N3O2
369.424
MFCD01863585
HLHBIMJNCKZZQO-SFTDATJTSA-N
2,6-bis 4r-4-phenyl-2-oxazolinyl pyridine,r,r-ph-pybox,2,6-bis r-4-phenyl-4,5-dihydrooxazol-2-yl pyridine,+-2,6-bis 4r-4-phenyl-2-oxazolin-2-yl pyridine,ph-pybox, r,r,2,6-bis 4r-phenyl-2-oxazolin-2-yl pyridine,r,r-2,6-bis 4-phenyl-2-oxazolin-2-yl pyridine,unii-555pn397nx,r,r-2,2'-2,6-pyridinediyl bis 4-phenyl-2-oxazoline,2,6-bis 4r-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl pyridine
2734679
(4R)-4-phenyl-2-[6-[(4R)-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole
C1C(N=C(O1)C2=NC(=CC=C2)C3=NC(CO3)C4=CC=CC=C4)C5=CC=CC=C5
Quantity 2g
Infrared Spectrum Authentic
Color White
Packaging Glass bottle
Melting Point 171°C to 174°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Merck Index 15, 8083
Formula Weight 369.42
Physical Form Needles
Percent Purity ≥97.5%
Chemical Name or Material 2, 6-Bis[(4R)-phenyl-2-oxazolin-2-yl]pyridine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.