Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Bis[(4S)-phenyl-2-oxazolin-2-yl]pyridine, Acros Organics

Click to view available options
Quantity:
2g
174500-20-0
C23H19N3O2
369.424
MFCD01863586
HLHBIMJNCKZZQO-NHCUHLMSSA-N
2,6-bis s-4-phenyl-4,5-dihydrooxazol-2-yl pyridine,2,6-bis 4s-phenyl-2-oxazolin-2-yl pyridine,s,s-ph-pybox,2,6-bis 4s-4-phenyl-2-oxazolinyl pyridine,--2,6-bis 4s-4-phenyl-2-oxazolin-2-yl pyridine,ph-pybox, s,s,unii-5r62cxu58k,s,s-2,6-bis 4-phenyl-2-oxazolin-2-yl pyridine,pyridine, 2,6-bis 4s-4,5-dihydro-4-phenyl-2-oxazolyl,2,6-bis 4s-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl pyridine
7010100
(4S)-4-phenyl-2-[6-[(4S)-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole
C1C(N=C(O1)C2=NC(=CC=C2)C3=NC(CO3)C4=CC=CC=C4)C5=CC=CC=C5
174500-20-0
C23H19N3O2
369.424
MFCD01863586
HLHBIMJNCKZZQO-NHCUHLMSSA-N
2,6-bis s-4-phenyl-4,5-dihydrooxazol-2-yl pyridine,2,6-bis 4s-phenyl-2-oxazolin-2-yl pyridine,s,s-ph-pybox,2,6-bis 4s-4-phenyl-2-oxazolinyl pyridine,--2,6-bis 4s-4-phenyl-2-oxazolin-2-yl pyridine,ph-pybox, s,s,unii-5r62cxu58k,s,s-2,6-bis 4-phenyl-2-oxazolin-2-yl pyridine,pyridine, 2,6-bis 4s-4,5-dihydro-4-phenyl-2-oxazolyl,2,6-bis 4s-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl pyridine
7010100
(4S)-4-phenyl-2-[6-[(4S)-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole
C1C(N=C(O1)C2=NC(=CC=C2)C3=NC(CO3)C4=CC=CC=C4)C5=CC=CC=C5
Quantity 2g
Merck Index 15, 8083
Formula Weight 369.42
Chemical Name or Material 2, 6-Bis[(4S)-phenyl-2-oxazolin-2-yl]pyridine