Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(6-Bromonaphthyl)-β-D-xylopyranoside hydrate, Acros Organics

Click to view available options
Quantity:
100mg
500mg
Packaging:
Glass bottle
69594-75-8
C15H15BrO5
355.184
MFCD04039743
WHFCYXMRDUCMTI-BARDWOONSA-N
6-bromo-2-naphthyl b-d-xylopyranoside,6-bromo-2-naphthyl beta-d-xylopyranoside,2-6-bromonaphthyl-beta-d-xylopyranoside,b-d-xylopyranoside,6-bromo-2-naphthalenyl 9ci,2s,3r,4s,5r-2-6-bromonaphthalen-2-yl oxy oxane-3,4,5-triol,6-bromo-2-naphthalenyl beta-d-xylopyranoside,2s,3r,4s,5r-2-6-bromonaphthalen-2-yl oxyoxane-3,4,5-triol,2s,3r,4s,5r-2-6-bromonaphthalen-2-yl oxy tetrahydro-2h-pyran-3,4,5-triol,2s,4s,3r,5r-2-6-bromo 2-naphthyloxy-2h-3,4,5,6-tetrahydropyran-3,4,5-trio l
2733919
(2S,3R,4S,5R)-2-(6-bromonaphthalen-2-yl)oxyoxane-3,4,5-triol
C1C(C(C(C(O1)OC2=CC3=C(C=C2)C=C(C=C3)Br)O)O)O
69594-75-8
C15H15BrO5
355.184
MFCD04039743
WHFCYXMRDUCMTI-BARDWOONSA-N
6-bromo-2-naphthyl b-d-xylopyranoside,6-bromo-2-naphthyl beta-d-xylopyranoside,2-6-bromonaphthyl-beta-d-xylopyranoside,b-d-xylopyranoside,6-bromo-2-naphthalenyl 9ci,2s,3r,4s,5r-2-6-bromonaphthalen-2-yl oxy oxane-3,4,5-triol,6-bromo-2-naphthalenyl beta-d-xylopyranoside,2s,3r,4s,5r-2-6-bromonaphthalen-2-yl oxyoxane-3,4,5-triol,2s,3r,4s,5r-2-6-bromonaphthalen-2-yl oxy tetrahydro-2h-pyran-3,4,5-triol,2s,4s,3r,5r-2-6-bromo 2-naphthyloxy-2h-3,4,5,6-tetrahydropyran-3,4,5-trio l
2733919
(2S,3R,4S,5R)-2-(6-bromonaphthalen-2-yl)oxyoxane-3,4,5-triol
C1C(C(C(C(O1)OC2=CC3=C(C=C2)C=C(C=C3)Br)O)O)O
Infrared Spectrum Authentic
Appearance (1% acetone/water 1:1) clear, colorless
Quantity 500mg
Color White
Melting Point 190°C to 196°C
Packaging Glass bottle
Water 5% Max. (K.F.)
Formula Weight 355.19
Physical Form Powder
Percent Purity Passes Test
Chemical Name or Material 2-(6-Bromonaphthyl)-β-D-xylopyranoside hydrate
Show More Show Less

Recommended Storage : Refrigerator +4°C